Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Trisodium hexanitritocobaltateSodium cobaltnitrite

Management of Change (MOC) category = N

BAKR3656-01 BAKR3656-01 BAKR3656-04
JT3656-1CS 7515.76 USD InStock
CAJT3656-1 JT3656-1 JT3656-4
Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Sodium hexanitritocobaltate(III)
For determination of potassium.

  • Lot analysis on label


Formula: Na₃Co(NO₂)₆
MW: 403.94 g/mol
Melting Pt: ∼220 °C
Storage Temperature: Ambient
MDL Number: MFCD00003512
CAS Number: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1,II
Merck Index: 12,08748
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For R&D use only. Not on TSCA Inventory.
Insoluble Matter ≤0.02 %
Suitability for Potassium Determination Passes Test
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty