Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium cyanide ≥95%, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKR3662-05 3662-01 BAKR3662-05
JT3662-1EA 408.39 USD InStock
JT3662-1 JT3662-5 CAJT3662-5
Sodium cyanide ≥95%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Sodium cyanide


Formula: CNNa
MW: 49.01 g/mol
Boiling Pt: 1496 °C (1013 hPa)
Melting Pt: 563.9 °C
Density: 1.595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1,I
Merck Index: 12,08750
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NaCN) (by Ag titrn) ≥ 95.0 %
Chloride (Cl) ≤ 0.15 %
Phosphate (PO₄) ≤ 0.02 %
Sulfate (SO₄) ≤ 0.05 %
Sulfide (S) ≤ 0.005 %
Thiocyanate (SCN) ≤ 0.02 %
Iron (Fe) ≤ 0.005 %
Trace Impurities - Lead (Pb) ≤ 5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty