Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dichromate dihydrate 99.5-100.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Danger
Synonyms: Sodium bichromate

Management of Change (MOC) category = N

BAKR3672-05 BAKR3672-01 BAKR3672-04
JT3672-1CS 1059.17 USD InStock
JT3672-1 JT3672-4 JT3672-5
Sodium dichromate dihydrate 99.5-100.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium dichromate dihydrate
  • Lot analysis on label


Formula: Na₂Cr₂O₇·2H₂O
MW: 298.00 g/mol (261.97 anhy)
Melting Pt: 356.7 °C
Density: 2.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II
Merck Index: 12,08754
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂Cr₂O₇·2H₂O) 99.5 - 100.5 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.005 %
Sulfate (SO₄) ≤0.01 %
Calcium (Ca) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Aluminum (Al) ≤0.0020 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty