Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

J.T. Baker®
BAKR3717-01 BAKR3717-07 BAKR3717-01 BAKR3717-07
JT3717-01EA 139.22 USD InStock
CAJT3717-01 JT3717-01 CAJT3717-07 JT3717-07
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
Meets ChP Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Insoluble Substances and Organic Matter Passes Test
NF - Potassium (K) ≤0.5 %
NF - Carbonate (as Na₂CO₃) ≤3.0 %
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
ChP -Assay 97.0 - 100.5 %
ChP -Sodium carbonate 2.0% max
ChP -Identification Passes Test
ChP -Alkalinity Passes Test
ChP -Clarity and color of solution  Passes Test
ChP -Chloride  0.005 % max
ChP -Sulfate  0.015 % max
ChP -Potassium  Passes Test
ChP -Aluminum and iron   0.1% max
ChP -Iron  0.001% max
ChP -Heavy metal  20 ppm max
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Carbonate (as Na₂CO₃) ≤2.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Assay (NaOH) 97.0 - 100.5 %
JP - Assay 95.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.050 %
JP - Heavy Metals (as Pb) ≤30 ppm
JP - Potassium (K) Passes Test
JP - Carbonate (as Na₂CO₃) ≤2.0 %
JP - Mercury (Hg) Passes Test
Endotoxin Concentration (EU/g) ≤5
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty