Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = N
J.T. Baker®
BAKR3727-25
BAKR3727-03
BAKR3727-09
BAKR3727-11
BAKR3727-07
BAKR3727-03
BAKR3727-R
BAKR3727-07
BAKR3727-01
BAKR3727-25
BAKR3727-R
BAKR3727-01
BAKR3727-09
BAKR3727-11
JT3727-25CS
12488.37
USD
InStock
JT3727-25
CAJT3727-11
CAJT3727-25
JT3727-11
JT3727-R
JT3727-3
JT3727-7
CAJT3727-1
CAJT3727-R
CAJT3727-3
JT3727-9
CAJT3727-7
CAJT3727-9
JT3727-1
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Sodium hydroxide
Caution: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.