Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 50% (by acidimetry), BAKER ANALYZED®, Electronic Grade, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = R

BAKR3735-07 3735-07
JT3735-7EA 159.89 USD InStock
JT3735-7 CAJT3735-7
Sodium hydroxide 50% (by acidimetry), BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
For Microelectronic Use
Protect from air to avoid absorption of carbon dioxide.
Store above 54 °F (12 °C) to avoid freezing.
Assay (NaOH) (by acidimetry) 49.5 - 51.5 %
Color (APHA) ≤10
Insoluble Matter ≤30 ppm
Sodium Carbonate (Na₂CO₃) ≤0.05 %
Ammonium Hydroxide Precipitate ≤0.007 %
Potassium (K) ≤0.005 %
Chloride (Cl) ≤20 ppm
Nitrogen Compounds (as N) ≤5 ppm
Phosphate (PO₄) ≤5 ppm
Sulfate (SO₄) ≤10 ppm
Heavy Metals (as Ag) ≤10 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Nickel (Ni) ≤3.000 ppm
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty