Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium metaperiodate

Management of Change (MOC) category = N

3756-04 BAKR3756-04 BAKR3756-01 BAKR3756-01
JT3756-1EA 1417.08 USD InStock
CAJT3756-1 CAJT3756-4 JT3756-1 JT3756-4
Sodium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Sodium periodate
  • Lot analysis on label


Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 175 °C
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 12,08786
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NaIO₄) (by iodometry) 99.8 - 100.3 %
Other Halogens (as Cl) ≤0.02 %
Trace Impurities - Manganese (Mn) ≤3.000 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty