Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium molybdenum oxide dihydrateMolybdic acid sodium salt dihydrateSodium molybdate dihydrateMolybdic acid, sodium salt dihydratedi-Sodium molybdate dihydrateMolybdic acid disodium salt dihydrate

Management of Change (MOC) category = N

BAKR3764-05 BAKR3764-R BAKR3764-01 BAKR3764-05 BAKR3764-01
JT3764-REA 17651.81 USD InStock
CAJT3764-5 JT3764-R JT3764-1 CAJT3764-1 JT3764-5
Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium molybdate(VI) dihydrate


Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Melting Pt: 687 °C
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (Na₂MoO₄·2H₂O) 99.5 - 103.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 7.0 - 10.5
Chloride (Cl) ≤0.005 %
Ammonium (NH₄) ≤0.001 %
Sulfate (SO₄) ≤0.015 %
Trace Impurities - Iron (Fe) ≤0.001 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty