Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Sodium Phosphate Monobasic hydrateSodium dihydrogen orthophosphate monohydrateSodium dihydrogenorthophosphate monohydrateSodium phosphate monobasic monohydrateSodium dihydrogen orthophosphate 1-hydrateSodium orthophosphate monobasic monohydrateMonosodium phosphateSodium dihydrogen phosphate 1-hydrate

Management of Change (MOC) category = NI

BAKR3818-09 BAKR3818-07 BAKR3818-R BAKR3818-09 BAKR3818-01 BAKR3818-05 BAKR3818-07 BAKR3818-01 BAKR3818-R BAKR3818-05
JT3818-09CS 1535.52 USD InStock
CAJT3818-7 CAJT3818-5 JT3818-09 JT3818-7 CAJT3818-1 CAJT3818-09 CAJT3818-R JT3818-5 JT3818-R JT3818-1
Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium dihydrogen phosphate monohydrate


Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/57/16640557.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NaH₂PO₄·H₂O) 98.0 - 102.0 %
pH of 5% Solution at 25°C 4.1 - 4.5
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
Potassium (K) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty