Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Sodium Phosphate Monobasic hydrateSodium dihydrogen orthophosphate monohydrateSodium dihydrogenorthophosphate monohydrateSodium phosphate monobasic monohydrateSodium dihydrogen orthophosphate 1-hydrateSodium orthophosphate monobasic monohydrateSodium dihydrogen phosphate 1-hydrate

Management of Change (MOC) category = HR

BAKR3820-09 BAKR3820-01 BAKR3820-R BAKR3820-R BAKR3820-07 BAKR3820-07 BAKR3820-09 BAKR3820-QM BAKR3820-QM BAKR3820-01
JT3820-7EA 2954.91 USD InStock
JT3820-7 CAJT3820-QM CAJT3820-1 JT3820-QM CAJT3820-09 JT3820-R JT3820-09 JT3820-1 CAJT3820-7 CAJT3820-R
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC, J.T.Baker®
Sodium dihydrogen phosphate monohydrate
  • Food GMP manufactured product


Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 12,08806
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/57/16640557.jpg
Order Now
Specification Test Results
Meets U.S.P Requirements
GMP Manufactured Product
Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
FCC - Assay (anhydrous basis) 98.0 - 103.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Fluoride (F) ≤0.005 %
FCC - Insoluble Substances ≤0.2 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 10.0 - 15.0 %
Appearance (White crystals or granules) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty