Synonyms:
Sodium Phosphate Monobasic hydrate
, Sodium dihydrogen orthophosphate monohydrate
, Sodium dihydrogenorthophosphate monohydrate
, Sodium phosphate monobasic monohydrate
, Sodium dihydrogen orthophosphate 1-hydrate
, Sodium orthophosphate monobasic monohydrate
, Sodium dihydrogen phosphate 1-hydrate
Management of Change (MOC) category = HR
BAKR3820-09
BAKR3820-01
BAKR3820-R
BAKR3820-R
BAKR3820-07
BAKR3820-07
BAKR3820-09
BAKR3820-QM
BAKR3820-QM
BAKR3820-01
JT3820-7EA
2954.91
USD
InStock
JT3820-7
CAJT3820-QM
CAJT3820-1
JT3820-QM
CAJT3820-09
JT3820-R
JT3820-09
JT3820-1
CAJT3820-7
CAJT3820-R
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC, J.T.Baker®
Sodium dihydrogen phosphate monohydrate
- Food GMP manufactured product