Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium monohydrogen phosphate dodecahydrate Sodium phosphate dibasic dodecahydrate di-Sodium hydrogen orthophosphate dodecahydrate

Management of Change (MOC) category = N

BAKR3822-01 BAKR3822-05 BAKR3822-07 BAKR3822-07 BAKR3822-01 BAKR3822-05
JT3822-5CS 1551.68 USD InStock
JT3822-5 JT3822-1 CAJT3822-7 CAJT3822-5 CAJT3822-1 JT3822-7
Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®
di-Sodium hydrogen phosphate dodecahydrate


Formula: Na₂HPO₄·12H₂O
MW: 358.14 g/mol
Melting Pt: 35 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay 98.0 - 102.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 8.7 - 9.3
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Nitrogen Compounds (as N) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty