Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogen phosphate heptahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: di-Sodium hydrogen phosphate 7-hydrateDisodium hydrogenorthophosphate 7-hydrateDisodium hydrogenorthophosphate heptahydrateDisodium phosphate (heptahydrate)Sodium monohydrogen phosphate 7-hydrateSodium monohydrogen phosphate heptahydrateSodium phosphate dibasic 7-hydrateSodium phosphate dibasic heptahydrate

Management of Change (MOC) category = NI

BAKR3824-07 BAKR3824-01 BAKR3824-05 3824-R BAKR3824-07 BAKR3824-01 BAKR3824-09 BAKR3824-09 BAKR3824-R BAKR3824-05
JT3824-REA 2669.93 USD InStock
JT3824-R JT3824-1 JT3824-09 CAJT3824-7 CAJT3824-09 CAJT3824-5 CAJT3824-R CAJT3824-1 JT3824-7 JT3824-5
Disodium hydrogen phosphate heptahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
di-Sodium hydrogen phosphate heptahydrate


Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48 °C
Density: 1.70 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂HPO₄·7H₂O) 98.0 - 102.0 %
pH of 5% Solution at 25°C 8.7 - 9.3
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty