Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium phosphate dibasic, anhydrous ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: di-Sodium hydrogen phosphatedi-Sodium hydrogen orthophosphate

BAKR3828-01 BAKR3828-07 BAKR3828-01 BAKR3828-05 BAKR3828-05 BAKR3828-09 BAKR3828-01 BAKR3828-R BAKR3828-07 BAKR3828-09 BAKR3828-R
JT3828-09CS 1666.06 USD InStock
CAJT3828-09 CAJT3828-7 JT3828-09 CAJT3828-5 CAJT3828-R CAJT3828-1 JT3828-7 JT3828-01 JT3828-R JT3828-05 JT3828-1
Sodium phosphate dibasic, anhydrous ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
di-Sodium hydrogen phosphate


Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂HPO₄) (by acidimetry) ≥99.0 %
Insoluble Matter ≤0.01 %
Loss on Drying at 105°C ≤0.2 %
pH of 5% Solution at 25°C 8.7 - 9.3
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.002 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty