Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (dry basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 3-(N-Morpholino)propanesulphonic acid3-Morpholinopropane-1-sulfonic acid3-Morpholinopropane-1-sulphonic acid3-(N-Morpholino)propanesulfonic acid

Management of Change (MOC) category = N

BAKR4004-01 4004-05 4004-01 BAKR4004-05
JT4004-1EA 589.47 USD InStock
CAJT4004-5 JT4004-1 JT4004-5 CAJT4004-1
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (dry basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®
γ-(N-Morpholino)propanesulphonic acid (MOPS)
DNase-, RNase-, and protease-free. Actual lot analysis reported on label.


Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 12,06346
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₇H₁₅NO₄S) (dried basis) ≥99 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
Appearance (white to slightly pink powder to crystalline powder) Passes Test
pH of 1% Solution at 25°C 2.5 - 4.5
Solution (10%) in Water Passes Test
Loss on Drying ≤1 %
Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm ≤0.020
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
pKa at 20 degrees C (typical) = 7.20

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty