Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium hydrogen phosphate ≥99.0%, powder, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: di-Potassium hydrogen orthophosphateDipotassium hydrogenphosphateDibasic Potassium PhosphatePotassium phosphate dibasicPotassium Phosphate, Dibasic, (dipotassium hydrogen phosphate)Potassium hydrogen phosphateDipotassium hydrogen phosphateDipotassium PhosphateDipotassium phosphate

Management of Change (MOC) category = NI

BAKR4012-05 BAKR4012-01 BAKR4012-05 BAKR4012-01
JT4012-1EA 322.4 USD InStock
JT4012-1 JT4012-5 CAJT4012-1 CAJT4012-5
di-Potassium hydrogen phosphate ≥99.0%, powder, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
di-Potassium hydrogen phosphate
Powder. For LC and molecular biology. Lot analysis on label.


Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 12,07828
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/42/16640642.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (K₂HPO₄) ≥99.0 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
pH of 5% Solution at 25°C 8.5 - 9.6
Chloride (Cl) ≤0.001 %
Insoluble Matter ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Loss on Drying at 105°C ≤1.0 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty