Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, crystalline, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 2-Morpholinoethanesulphonic acid hydrate2-(N-Morpholino)ethanesulfonic acid monohydrate2-(N-Morpholino)ethanesulphonic acid monohydrate2-Morpholinoethanesulfonic acid hydrate

Management of Change (MOC) category = N

BAKR4014-07 4014-01 BAKR4014-04 BAKR4014-01 BAKR4014-02 BAKR4014-04 BAKR4014-02 BAKR4014-07
JT4014-1EA 733.38 USD InStock
CAJT4014-7 JT4014-1 JT4014-2 JT4014-4 CAJT4014-1 JT4014-7 CAJT4014-4 CAJT4014-2
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, crystalline, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Buffer for liquid chromatography and other molecular biology applications. Lot analysis on label.


Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₃NO₄S·H₂O) ≥98 %
Appearance Passes Test
Loss on Drying at 130°C 7 - 10 %
Heavy Metals (as Pb) ≤2 ppm
Product Information (not a specification):
pKa at 20 degrees C = 6.15

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty