Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formamide ≥99.5% (by GC), Ultrapure Bioreagent for denaturing nucleic acids prior to analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Carbamaldehyde

Management of Change (MOC) category = N

BAKR4028-00 BAKR4028-01 BAKR4028-01 BAKR4028-00
JT4028-0EA 113.23 USD InStock
JT4028-0 JT4028-1 CAJT4028-1 CAJT4028-0
Formamide ≥99.5% (by GC), Ultrapure Bioreagent for denaturing nucleic acids prior to analysis, J.T.Baker®
Formamide
For denaturing nucleic acids prior to analysis.

ULTRAPURE BIOREAGENT—A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.


Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (HCONH₂) (by GC) ≥99.5 %
Absorbance of a 1M Solution - 280 nm ≤0.100
Absorbance of a 1M Solution - Color (APHA) ≤10
Absorbance of a 1M Solution - Conductivity, mS/cm (1:10), For Information Only
Freezing Point 2.0 - 3.0 °C
Trace Impurities - Copper (Cu) ≤0.1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Trace Impurities - Lead (Pb) ≤0.5 ppm
Trace Impurities - Zinc (Zn) ≤0.5 ppm
Packaged under Nitrogen
Deionization of material is not required.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty