Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Aminoacetic acidH-Gly-OH

Management of Change (MOC) category = N

BAKR4059-07 BAKR4059-07 BAKR4059-02 BAKR4059-00 BAKR4059-00 BAKR4059-06 BAKR4059-06 BAKR4059-02
JT4059-2EA 205.39 USD InStock
JT4059-2 CAJT4059-6 CAJT4059-7 JT4059-0 JT4059-6 CAJT4059-2 JT4059-7 CAJT4059-0
Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®
Glycine


Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration) ≥99.5 %
Homogeneity by TLC Passes Test
Residue after Ignition ≤0.10 %
Loss on Drying at 105°C ≤0.2 %
Readily Carbonizable Substances Passes Test
Absorbance of a 1M Solution - 280 nm ≤0.100
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Arsenic (As) ≤3 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤0.003 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty