Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Sucrose, crystals NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: alpha-D-Glucopyranosidebeta-D-Fructofuranosyl alpha-D-glucopyranosideD-(+)-SaccharoseSucroseSugar

Management of Change (MOC) category = R

J.T. Baker®
BAKR4074-20 BAKR4074-01 BAKR4074-05 BAKR4074-20 BAKR4074-05 BAKR4074-07 BAKR4074-19 BAKR4074-19 BAKR4074-QM BAKR4074-01 BAKR4074-07 BAKR4074-QM
JT4074-QMCS 29.1 USD InStock
CAJT4074-1 JT4074-QM JT4074-7 CAJT4074-19 JT4074-19 JT4074-5 JT4074-1 CAJT4074-5 CAJT4074-20 JT4074-20 CAJT4074-7 CAJT4074-QM
D-(+)-Sucrose, crystals NF, Multi-Compendial, J.T.Baker®
D(+)-Sucrose


Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.59 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 12,09051
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg
Order Now
Specification Test Results
GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Appearance of Solution Passes Test
NF - Color Value ≤45
NF - Conductivity ≤35 µS/cm
NF - Optical Rotation (+) 66.3 - 67.0 Degree
NF - Reducing Sugars Passes Test
NF - Sulfite (SO2) ≤10 ppm
NF - Loss on Drying at 105 ºC ≤0.1 %
EP/BP - Identification A (EP/BP) Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤35
EP/BP - Optical Rotation (+) 66.3 - 67.0 Degree
EP/BP - Colour Value ≤45
EP/BP - Reducing Sugars Passes Test
EP/BP - Sulfite (SO₂) ≤10 ppm
EP/BP - Loss on Drying at 105°C ≤0.1 %
JP - Identification Passes Test
JP - Optical Rotation (+) 66.3 - 67.0 Degree
JP - Clarity of Solution Passes Test
JP - Color Value ≤45
JP - Sulfite (SO₂) ≤10 ppm
JP - Reducing Sugars Passes Test
JP - Conductivity, uS cm-1 ≤35
JP - Loss on Drying at 105°C ≤0.1 %
Endotoxin Concentration (EU/g) ≤250
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Store in a tightly closed container.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty