Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Tris(hydroxymethyl)aminomethane hydrochlorideTris(hydroxymethyl)methylamine hydrochlorideTrizma® hydrochlorideTromethane hydrochlorideTrizma(R) hydrochlorideTris(hydroxymethyl)methylammonium chlorideTHAM HClTris-HClTrometamol hydrochloride2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochlorideTRIS HCl

Management of Change (MOC) category = N

BAKR4103-06 BAKR4103-F7 BAKR4103-07 BAKR4103-F7 BAKR4103-01 BAKR4103-07 4103-04 BAKR4103-01 BAKR4103-04 BAKR4103-02 BAKR4103-08 BAKR4103-06 BAKR4103-08 BAKR4103-02
JT4103-08CS 15764.57 USD InStock
JT4103-08 JT4103-2 JT4103-1 CAJT4103-1 CAJT4103-2 CAJT4103-4 CAJT4103-6 CAJT4103-7 JT4103-7 CAJT4103-8 JT4103-6 JT4103-4 JT4103-F7 CAJT4103-F7
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
For liquid chromatography and molecular biology applications.

  • Lot analysis on label


Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 150…152 °C
Density: 1.05 g/ml (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/01/2297601.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (NH₂C(CH₂OH)₃·HCl) (dried basis) ≥99.0 %
Appearance Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Trace Impurities - Calcium (Ca) ≤1 ppm
Trace Impurities - Copper (Cu) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤1 ppm
Trace Impurities - Magnesium (Mg) ≤1.000 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty