Synonyms:
Tris(hydroxymethyl)aminomethane hydrochloride
, Tris(hydroxymethyl)methylamine hydrochloride
, Trizma(R) hydrochloride
, Tris(hydroxymethyl)methylammonium chloride
, THAM HCl
, Trometamol hydrochloride
, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride
, TRIS HCl
Management of Change (MOC) category = N
BAKR4103-06
BAKR4103-F7
BAKR4103-07
BAKR4103-F7
BAKR4103-01
BAKR4103-07
4103-04
BAKR4103-01
BAKR4103-04
BAKR4103-02
BAKR4103-08
BAKR4103-06
BAKR4103-08
BAKR4103-02
JT4103-08CS
15764.57
USD
InStock
JT4103-08
JT4103-2
JT4103-1
CAJT4103-1
CAJT4103-2
CAJT4103-4
CAJT4103-6
CAJT4103-7
JT4103-7
CAJT4103-8
JT4103-6
JT4103-4
JT4103-F7
CAJT4103-F7
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
For liquid chromatography and molecular biology applications.