Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9% (dried basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Trizma(R) baseTRIS baseTHAM2-Amino-2-(hydroxymethyl)-1,3-propanediolTris(hydroxymethyl)methylamine2-Amino-2-(hydroxymethyl)propane-1,3-diol2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris)Tromethamine

Management of Change (MOC) category = NI

BAKR4109-09 BAKR4109-07 BAKR4109-F7 BAKR4109-01 BAKR4109-06 BAKR4109-F7 BAKR4109-02 BAKR4109-09 BAKR4109-06 BAKR4109-07 BAKR4109-01 BAKR4109-02
JT4109-2EA 367.31 USD InStock
CAJT4109-1 JT4109-2 JT4109-1 CAJT4109-2 JT4109-F7 CAJT4109-6 CAJT4109-7 CAJT4109-9 CAJT4109-F7 JT4109-09 JT4109-7 JT4109-6
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9% (dried basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Buffer for molecular biology and liquid chromatography.

  • Lot analysis on label

Material also meets the chemical specifications of Tromethamine, USP.


Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 12,09902
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/00/2297600.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₄H₁₁NO₃) (dried basis) ≥99.9 %
Assay (C₄H₁₁NO₃) 99.0 - 101.0 %
Appearance (White crystals) Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
pH of 0.1M Solution at 25°C 10.0 - 11.0
Insoluble Matter ≤0.005 %
Water (H₂O)(by Karl Fischer titrn) ≤0.3 %
Trace Impurities - Arsenic (As) ≤1 ppm
Trace Impurities - Calcium (Ca) ≤1 ppm
Trace Impurities - Copper (Cu) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤1 ppm
Trace Impurities - Magnesium (Mg) ≤1.000 ppm
Trace Impurities - Manganese (Mn) ≤1 ppm
Identification A Passes Test
Identification B Passes Test
Identification C Passes Test
Description Passes Test
pH (1 in 20) 10.0 - 11.5
Loss on Drying ≤1.0 %
Residue after Ignition ≤0.10 %
Heavy Metals (as Pb) ≤0.001 %
Melting Range(Lower) 168 - 172 °C
Melting Range(Upper) 168 - 172 °C
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Material also meets the chemical specifications of Tromethamine, U.S.P.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty