Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HEPES sodium salt ≥99.0% (dried basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium saltSodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonateSodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonateSodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonateSodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium saltN-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

Management of Change (MOC) category = N

4153-06 BAKR4153-01 BAKR4153-05 BAKR4153-01 BAKR4153-05 BAKR4153-06
JT4153-1CS 669.86 USD InStock
JT4153-1 JT4153-06 JT4153-5 CAJT4153-1 CAJT4153-5 CAJT4153-6
HEPES sodium salt ≥99.0% (dried basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®
HEPES sodium salt
Ultrapure Bioreagent - A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.


Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Melting Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/70/18366170.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₈H₁₇N₂NaO₄S) (dried basis) ≥99.0 %
Appearance Passes Test
Solubility Passes Test
Loss on Drying at 80°C ≤3.0 %
Insoluble Matter ≤1.0 %
Heavy Metals (as Pb) ≤1 ppm
Identification (Sodium) Passes Test
pH of 5% Solution at 25°C 9.5 - 11.5
pK^a at 20°C, For Information Only
QC Supervisor or equivalent to check supplier's COA to ensure proper chemical form. Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty