Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 2-Morpholinoethanesulphonic acid hydrate2-(N-Morpholino)ethanesulfonic acid monohydrate2-(N-Morpholino)ethanesulphonic acid monohydrate2-Morpholinoethanesulfonic acid hydrate

Management of Change (MOC) category = R

BAKR4220-06 BAKR4220-08 BAKR4220-08 BAKR4220-06
JT4220-06CS 17815.38 USD InStock
JT4220-06 CAJT4220-06 JT4220-08 CAJT4220-08
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Suitable for use in biopharmaceutical manufacturing applications.


Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
MDL Number: MFCD00149409
CAS Number: 145224-94-8
Order Now
Specification Test Results
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (C6H13NO4S·H2O) ≥99.0 %
Appearance and Color (White crystals) Passes Test
Identification (by IR) Passes Test
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Color of a 1M Alkaline Solution (Colorless) Passes Test
Loss on Drying at 130°C 7 - 10 %
Residue on Ignition ≤0.05 %
Heavy Metals (as Pb) ≤2 ppm
Absorbance, 1M Solution (au): 260 nm ≤0.1000
Absorbance, 1M Solution (au): 280 nm ≤0.1000
Chloride (Cl) ≤0.005 %
pH (5% solution) 3.1 - 3.5
pH (0.5M ) 2.5 - 4.5
pKa 5.9 - 6.3
Solubility Passes Test
Sulfate (SO4) ≤0.005 %
Arsenic (As) ≤5 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty