Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc acetate dihydrate 98.0-102.0%, crystals, Technical Grade, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Zinc diacetate dihydrateAcetic acid zinc salt dihydrateZinc (II) acetate dihydrate

Management of Change (MOC) category = N

BAKR4304-R BAKR4304-R
JT4304-REA 4887.1 USD InStock
JT4304-R CAJT4304-R
Zinc acetate dihydrate 98.0-102.0%, crystals, Technical Grade, J.T.Baker®
Zinc acetate dihydrate


Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃COO)₂Zn·2H₂O) 98.0 - 102.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 6.0 - 7.0
Lead (Pb) ≤0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty