Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc oxide ≥99.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Zinci oxidumZinc (II) oxide

Management of Change (MOC) category = N

BAKR4358-05 BAKR4358-R BAKR4358-01 BAKR4358-01 BAKR4358-R BAKR4358-05
JT4358-5CS 1570.58 USD InStock
CAJT4358-1 CAJT4358-R JT4358-5 JT4358-1 JT4358-R CAJT4358-5
Zinc oxide ≥99.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Zinc oxide


Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (ZnO) ≥99.0 %
ACS - Insoluble in Dilute H₂SO₄ ≤0.01 %
ACS - Alkalinity Passes Test
ACS - Nitrate (NO₃) ≤0.003 %
ACS - Sulfur Compounds (as SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Lead (Pb) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤5.0 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty