Would you like to log in again?
J.T.Baker® BAKER-phix, pH Indicator Sticks
Supplier: Avantor
BAKR4393-01 BAKR4392-01 BAKR4393-01 BAKR4390-01 BAKR4391-01 BAKR4395-01 BAKR4390-01 BAKR4392-01 BAKR4398-01 BAKR4393-01 BAKR4396-01 BAKR4397-01 BAKR4391-01 BAKR4397-01 BAKR4390-01
JT4393-1PK 40.21 USD InStock
JT4393-1 CAJT43971 JT4390-1 JT43901 JT43911 JT43981 JT43961 JT43971 JT43951 JT43921 JT43931 CAJT43911 CAJT43901 CAJT43931 CAJT43921
J.T.Baker® BAKER-phix, pH Indicator Sticks
Test Paper pH Indicator Papers and Test Strips
A range of pH indicator sticks that feature up to four color-fixed test zones on a durable plastic strip.
Made with color-fixed, non-bleeding dyes that won’t contaminate your samples, these indicator sticks contain a larger scale and well-separated dyes for easy readability. Solutions do not need to be buffered before testing.
Each stick measures ¼×3¼" (6×85 mm).
Delivery information: Packaged in easy-to-use plastic boxes containing 100 sticks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty