Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfate 99.0-100.5% NF, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Diammonium sulfateSulphuric acid diammonium saltDiammonium sulphateSulfuric acid diammonium salt

Management of Change (MOC) category = R

J.T. Baker®
BAKR4628-01 BAKR4628-65 BAKR4628-01 BAKR4628-F9 BAKR4628-F9
JT4628-F9CS 14189.92 USD InStock
CAJT4628-01 JT4628-F9 JT4628-01 JT4628-65 CAJT4628-F9
Ammonium sulfate 99.0-100.5% NF, ACS, J.T.Baker®
Ammonium sulphate
  • GMP manufactured product


Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
NF - Insoluble Matter ≤ 0.005 %
NF - Residue on Ignition ≤ 0.005 %
NF - pH of 5% Solution 5.0 - 6.0
NF - Nitrate (NO₃) ≤ 0.001 %
NF - Chloride (Cl) ≤ 5 ppm
NF - Phosphate (PO₄) ≤ 5 ppm
NF - Iron (Fe) ≤ 5 ppm
NF - Identification Passes Test
NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g) ≤ 1000
NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g) ≤ 10
Endotoxin Concentration (EU/g) ≤ 2.5
Nickel (Ni) ≤ 0.7 ppm
Cobalt (Co) ≤ 0.09 ppm
Chromium (Cr) ≤ 0.75 ppm
Copper (Cu) ≤ 0.03 ppm
Manganese (Mn) ≤ 3.45 ppm
Zinc (Zn) (For Information Only)
Ni / Zn Ratio (For Information Only)
Co / Zn Ratio (For Information Only)
Cu / Zn Ratio (For Information Only)
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥ 99.0 %
ACS - Residue on Ignition ≤ 0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Insoluble Matter ≤ 0.005 %
ACS - Chloride (Cl) ≤ 5 ppm
ACS - Nitrate (NO₃) ≤ 0.001 %
ACS - Phosphate (PO₄) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 5 ppm
ACS - Iron (Fe) ≤ 5 ppm
Solution Test Passes Test
No Class 1, 2 or 3 Residual Solvents are likely to be present by
exclusion of the production/purification process.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Preserve in well closed container.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty