Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

Management of Change (MOC) category = N

BAKR4681-01 BAKR4681-01
JT4681-1CS 3418.88 USD InStock
JT4681-1 CAJT4681-1
Silver nitrate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1760
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
For laboratory research or manufacturing use.
Normality (by titrimetry) Passes Test
(Makes 0.1N solution after dilution to 1000 ml)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty