Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = N

BAKR4689-01
JT4689-1CS 425.57 USD InStock
JT4689-1
Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide
(1 equiv.=40.00 g NaOH)


Formula: NaOH
MW: 40 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
For laboratory, research or manufacturing use.
Normality (by titrimetry) Passes Test
(Makes 1N solution after dilution to 1000 ml)
Low in carbonate

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty