Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 2-Morpholinoethanesulphonic acid hydrate2-(N-Morpholino)ethanesulfonic acid monohydrate2-(N-Morpholino)ethanesulphonic acid monohydrate2-Morpholinoethanesulfonic acid hydrate

Management of Change (MOC) category = NI

J.T. Baker®
BAKR4812-06 BAKR4812-02 BAKR4812-06 BAKR4812-08 BAKR4812-04 BAKR4812-02 BAKR4812-08
JT4812-6EA 14195.53 USD InStock
CAJT4812-08 CAJT4812-2 JT4812-6 CAJT4812-6 CAJT4812-4 JT4812-08 JT4812-2
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Suitable for use in biopharmaceutical manufacturing applications.


Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₃NO₄S·H₂O) ≥98 %
Appearance Passes Test
Identification (by IR) Passes Test
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Color of a 1M Alkaline Solution (Colorless) Passes Test
Loss on Drying at 130°C 7 - 10 %
Heavy Metals (as Pb) ≤2 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty