Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MES-Na ≥85.0% for biopharmaceutical production, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: MES-NaSodium 3-morpholin-1-ylethylsulfonate2-(N-Morpholino)ethanesulphonic acid sodium saltSodium 3-morpholin-1-ylethylsulphonate2-(N-Morpholino)ethanesulfonic acid sodium saltMES sodium salt

Management of Change (MOC) category = NI

BAKR4813-02 4813-04 BAKR4813-06 BAKR4813-02 4813-03 BAKR4813-02 BAKR4813-06 4813-01 BAKR4813-01 BAKR4813-02
JT4813-6EA 13869.13 USD InStock
CAJT4813-2 CAJT4813-1 JT4813-6 CAJT4813-6 CAJT4813-4 TXLJT4813-2SPH JT4813-3 CILJT4813-2SPH JT4813-1 JT4813-2
MES-Na ≥85.0% for biopharmaceutical production, purified, J.T.Baker®
MES sodium salt
Suitable for use in biopharmaceutical manufacturing applications.


Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
MDL Number: MFCD00065473
CAS Number: 71119-23-8
EINECS: 275-203-2
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₂NNaO₄S) ≥85.0 %
Appearance Passes Test
Identification (by IR) Passes Test
pH of 1% Aqueous Solution at 25°C 8.9 - 10.1
Solubility Passes Test
Ultraviolet Absorbance of a 10% Aqueous Solution - 280 nm ≤0.03
Ultraviolet Absorbance of a 10% Aqueous Solution - 260 nm ≤0.05
Water (H₂O) ≤15.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty