Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen phthalate 99.95-100.05% (dried basis), ULTREX®, Ultrapure, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: KHPPotassium phthalate monobasicPotassium Acid PhthalatePotassium biphthalatePhthalic Acid Monopotassium SaltPhthalic acid potassium salt

Management of Change (MOC) category = N

BAKR4889-03 BAKR4889-03
JT4889-3EA 813.52 USD InStock
CAJT4889-3 JT4889-3
Potassium hydrogen phthalate 99.95-100.05% (dried basis), ULTREX®, Ultrapure, J.T.Baker®
Potassium hydrogen phthalate
For primary standard and research applications.


Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Melting Pt: 295…300 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 12,07773
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/58/16640658.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (1-KOCOC₆H₄-2-COOH) (dried basis) 99.95 - 100.05 %
Identification (by IR) Passes Test
Particulate Matter, For Information Only
pH of 0.05M Solution at 25°C 4.00 - 4.02
Non-Metallic Impurities - Chlorine Compounds (as Cl), For Information Only
Non-Metallic Impurities - Silicon (Si), For Information Only
Non-Metallic Impurities - Sulfur Compounds (as S), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Copper (Cu), For Information Only
Metallic Impurities - Iron (Fe), For Information Only
Metallic Impurities - Lead (Pb), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Mercury (Hg), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Niobium (Nb), For Information Only
Metallic Impurities - Silver (Ag), For Information Only
Metallic Impurities - Sodium (Na), For Information Only
Metallic Impurities - Strontium (Sr), For Information Only
Metallic Impurities - Tin (Sn), For Information Only
Metallic Impurities - Titanium (Ti), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Metallic Impurities - Zirconium (Zr), For Information Only
Certificate Provided Reports Actual Lot Analysis

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty