Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = R

J.T. Baker®
BAKR5000-07 BAKR5000-02 BAKR5000-R BAKR5000-09 BAKR5000-27 BAKR5000-27 BAKR5000-30 BAKR5000-QM BAKR5000-R BAKR5000-30 BAKR5000-09 BAKR5000-QM BAKR5000-07 BAKR5000-02 5000-03 BAKR5000-03
JT5000-7EA 2547.16 USD InStock
JT5000-7 JT5000-R JT5000-3 CAJT5000-9 CAJT5000-7 CAJT5000-3 CAJT5000-R JT5000-02 CAJT5000-02 JT5000-27 CAJT5000-27 JT5000-QM JT5000-30 CAJT5000-30 JT5000-9 CAJT5000-QM
Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet Ph. Eur., BP, and JP chemical specifications.
Filtered through a 0.2 µm filter. Completely miscible with water. GMP manufactured product.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
GMP Manufactured Product
IMPORTANT: Material will freeze if stored below 41 °F (5°C).
Protect from air to avoid absorption of carbon dioxide.
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 9.9 - 10.1
Sodium Carbonate (Na₂CO₃) ≤0.6 %
Chloride (Cl) ≤20 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤2 ppm
Endotoxin Concentration (EU/mL) ≤2.0
Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
IMPORTANT: Material will freeze if stored below 41°F (5°C).
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty