Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = R
J.T. Baker®
BAKR5000-07
BAKR5000-02
BAKR5000-R
BAKR5000-09
BAKR5000-27
BAKR5000-27
BAKR5000-30
BAKR5000-QM
BAKR5000-R
BAKR5000-30
BAKR5000-09
BAKR5000-QM
BAKR5000-07
BAKR5000-02
5000-03
BAKR5000-03
JT5000-7EA
2547.16
USD
InStock
JT5000-7
JT5000-R
JT5000-3
CAJT5000-9
CAJT5000-7
CAJT5000-3
CAJT5000-R
JT5000-02
CAJT5000-02
JT5000-27
CAJT5000-27
JT5000-QM
JT5000-30
CAJT5000-30
JT5000-9
CAJT5000-QM
Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet Ph. Eur., BP, and JP chemical specifications.
Filtered through a 0.2 µm filter. Completely miscible with water. GMP manufactured product.