Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylene glycol ≥99% VLSI, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 1,2-Dihydroxyethane2-HydroxyethanolDihydroxyethaneEthanediolEthylene alcoholGlycol alcoholMonoethylene glycol

Management of Change (MOC) category = R

5387-03 BAKR5387-03 5387-05 BAKR5387-05
JT5387-3CS 1658.28 USD InStock
JT5387-3 JT5387-5 CAJT5387-5 CAJT5387-3
Ethylene glycol ≥99% VLSI, J.T.Baker®
Ethylene glycol


Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/734375.jpg
Order Now
Specification Test Results
For Microelectronic Use
Assay (HOCH₂CH₂OH) (by GC) ≥99.0 %
Color (APHA) ≤10
Acidity (µeq/g) ≤0.8
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤1 ppm
Arsenic and Antimony (as As) ≤0.100 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Aluminum (Al) ≤100.0 ppb
Trace Impurities - Barium (Ba) ≤100.0 ppb
Trace Impurities - Boron (B) ≤50 ppb
Trace Impurities - Calcium (Ca) ≤300 ppb
Trace Impurities - Chromium (Cr) ≤50 ppb
Trace Impurities - Copper (Cu) ≤50 ppb
Trace Impurities - Gold (Au) ≤50 ppb
Trace Impurities - Lead (Pb) ≤200.0 ppb
Trace Impurities - Lithium (Li) ≤100.0 ppb
Trace Impurities - Magnesium (Mg) ≤100.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Sodium (Na) ≤800.0 ppb
Trace Impurities - Tin (Sn) ≤100 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Zinc (Zn) ≤400.0 ppb
Particle Count - 0.5 µm and greater ≤130 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty