Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 97.5-100.5% (by acidimetry), pellets, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

BAKR5565-25 BAKR5565-28 BAKR5565-25 BAKR5565-28
JT5565-25CS 2570.3 USD InStock
JT5565-25 CAJT5565-25 CAJT5565-28 JT5565-28
Sodium hydroxide 97.5-100.5% (by acidimetry), pellets, J.T.Baker®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NaOH) (by acidimetry) 97.5 - 100.5 %
ACS - Sodium Carbonate (Na₂CO₃) ≤1.0 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Ag) ≤0.002 %
ACS - Potassium (K) ≤0.02 %
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.003 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Mercury (Hg) ≤0.100 ppm
ACS - Nickel (Ni) ≤0.001 %
EP/BP - Characteristics Passes Test
EP/BP - Identification Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Heavy Metals (as Pb) ≤20 ppm
EP/BP - Iron (Fe) ≤10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty