Would you like to log in again?
J.T.Baker®, pH Buffer Solutions
Supplier: Avantor
J.T.Baker®
BAKR5606-01 BAKR5655-03 BAKR5655-03 BAKR5606-03 BAKR5657-07 BAKR5655-01 BAKR5655-07 BAKR5656-07 BAKR5657-03 BAKR5657-07 BAKR5606-03 BAKR5655-01 BAKR5657-01 BAKR5657-03 BAKR5657-02 BAKR5608-01 BAKR5606-01 BAKR5608-03 BAKR5657-01 BAKR5656-01
40004-722CS 446.18 USD InStock
40004-722 CAJT5657-7 CAJT5606-1 CAJT5656-7 JT5655-1 CAJT5657-3 CAJT5606-3 CAJT5655-7 CAJT5655-1 CAJT5655-3 JT5657-7 CAJT5657-1 CAJT5656-1 JT5655-3 JT5657-1 JT5606-3 JT5608-1 JT5657-3 JT5606-1 JT5608-3
J.T.Baker®, pH Buffer Solutions
Buffers pH Calibration Buffers
High quality buffer solutions ensure accurate pH measurements time after time.
  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and color-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Certified traceable to NIST Standard Reference Buffers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty