Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

Management of Change (MOC) category = N

BAKR5614-03 BAKR5614-02 BAKR5614-03
JT5614-2CS 494.8 USD InStock
JT5614-2 JT5614-3 CAJT5614-3
Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Hydrochloric acid
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, provided on label.


Formula: HCl
MW: 36.46 g/mol
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Normality 0.0195 - 0.0205
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty