Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfiteAntichlorSodiumthiosulphate

Management of Change (MOC) category = NI

BAKR5637-03 BAKR5637-02 BAKR5637-02 BAKR5637-07 BAKR5637-03 BAKR5637-07
JT5637-2CS 395.11 USD InStock
JT5637-2 JT5637-3 CAJT5637-7 JT5637-7 CAJT5637-3 CAJT5637-2
Sodium thiosulfate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium thiosulphate


Formula: Na₂S₂O₃
MW: 158.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Sulfate and Sulfite (as SO₄) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤1 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty