Synonyms:
Ethylenediaminetetra-acetic acid disodium salt dihydrate
, Edetate disodium dihydrate
, Kestranal® 2S 2 H2O
, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate
, Titriplex III
, Edetic acid disodium salt dihydrate
, Kestranal® 2S dihydrate
, Disodium dihydrogen ethylenediaminetetraacetate dihydrate
Management of Change (MOC) category = NI
BAKR5648-03
BAKR5648-02
BAKR5648-03
BAKR5648-02
JT5648-2CS
551.46
USD
InStock
JT5648-2
JT5648-3
CAJT5648-3
CAJT5648-2
EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL), J.T.Baker®
EDTA disodium salt
Standardized at 25 °C against calcium carbonate whose certification is traceable to NIST.