Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIEdetic acid disodium salt dihydrateKestranal® 2S dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate

Management of Change (MOC) category = NI

BAKR5648-03 BAKR5648-02 BAKR5648-03 BAKR5648-02
JT5648-2CS 551.46 USD InStock
JT5648-2 JT5648-3 CAJT5648-3 CAJT5648-2
EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL), J.T.Baker®
EDTA disodium salt
Standardized at 25 °C against calcium carbonate whose certification is traceable to NIST.


Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Titer, mg/mL, For Information Only
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Each lot of this product is standardized at 25°C against NIST StandardReference Material Calcium Carbonate (Hydroxy Naphthol Blue endpoint).
SRM No…
The titer found falls within the range 0.980-1.020 mg CaCO₃/mL.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty