Synonyms:
Permanganic acid potassium salt
Management of Change (MOC) category = NI
BAKR5651-03
BAKR5651-02
BAKR5651-03
JT5651-3CS
504.43
USD
InStock
CAJT5651-3
JT5651-3
JT5651-2
Potassium permanganate 0.0995-0.1005 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Potassium permanganate
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.