Synonyms:
Caustic sodaLyeSodium hydrateSoda lye
Management of Change (MOC) category = NI
J.T. Baker®
BAKR5653-03
BAKR5653-03
BAKR5653-02
BAKR5653-02
JT5653-03CS
726.53
USD
InStock
CAJT5653-3
CAJT5653-2
JT5653-03
JT5653-2
Sodium hydroxide 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.