Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

Management of Change (MOC) category = NI

J.T. Baker®
BAKR5653-03 BAKR5653-03 BAKR5653-02 BAKR5653-02
JT5653-03CS 726.53 USD InStock
CAJT5653-3 CAJT5653-2 JT5653-03 JT5653-2
Sodium hydroxide 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.


Formula: NaOH
MW: 40 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
ADR: 8,III
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Normality 0.0195 - 0.0205
Chloride (Cl) ≤1 ppm
Trace Impurities - Heavy Metals ≤0.5 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty