Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = NI
BAKR5663-03
BAKR5663-02
BAKR5663-03
BAKR5663-02
JT5663-2CS
355.25
USD
InStock
CAJT5663-3
CAJT5663-2
JT5663-2
JT5663-3
Sodium hydroxide 0.01 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.