Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = NI
BAKR5665-03
BAKR5665-02
BAKR5665-07
BAKR5665-07
BAKR5665-02
JT5665-7EA
350.26
USD
InStock
CAJT5665-3
JT5665-7
CAJT5665-2
JT5665-2
CAJT5665-7
Sodium hydroxide 0.2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Standardization at 25 °C traceable to NIST Standard Reference Material.