Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = NI

J.T. Baker®
BAKR5674-17 BAKR5674-02 BAKR5674-17 BAKR5674-06 BAKR5674-09 BAKR5674-06 BAKR5674-07 BAKR5674-03 BAKR5674-03 BAKR5674-09
JT5674-17EA 391.46 USD InStock
JT5674-17 CAJT5674-6 CAJT5674-3 JT5674-09 CAJT5674-17 JT5674-6 JT5674-3 CAJT5674-09 JT5674-2 CAJT5674-7
Sodium hydroxide 10 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Caution: IMPORTANT: Material will freeze if stored below 41 °F (5 °C). Protect from air to avoid absorption of carbon dioxide.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
IMPORTANT: Material will freeze if stored below 41 °F (5°C).
Appearance (Clear, colorless liquid) Passes Test
Normality 9.9 - 10.1
Sodium Carbonate (Na₂CO₃) ≤2.0 %
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…
IMPORTANT: Material will freeze if stored below 41°F (5°C).

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty