Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-1-methylethyl acetate (PGMEA) ≥99.0% (by GC) stabilized (BTS-220), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: 1-Methoxy-2-propyl acetateDOWANOL PMA2-(1-Methoxy)propyl acetatePropylene glycol 1-methyl ether 2-acetatPropylene glycol methyl ether acetat1,2-Propanediol monomethyl ether acetate

6343-05 6343-05
JT6343-5CS 1121.2 USD InStock
JT6343-5 CAJT6343-5
2-Methoxy-1-methylethyl acetate (PGMEA) ≥99.0% (by GC) stabilized (BTS-220), J.T.Baker®
2-Methoxy-1-methylethyl acetate (PGMEA)
Edge bead remover (PGMEA).


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 145.8 °C (1013 hPa)
Melting Pt: –67 °C
Density: 0.968 g/cm³ (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00038500
CAS Number: 108-65-6
EINECS: 203-603-9
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Acidity (as CH3COOH) 0.02% max.
Appearance Pass test
Calcium (Ca) 20 ppb max.
Color (APHA) 10 max.
Iron (Fe) 20 ppb max.
Magnesium (Mg) 20 ppb max.
Potassium (K) 20 ppb max.
Sodium (Na) 20 ppb max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty